
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-1-ethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053656-91-9
:
Formula:C₈H₈ClF₃N₂O
MDL Number:MFCD10568236
:240.61 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together