3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide
Catalog # APOSPC300549-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.7 g/mol
Storage Temperature:Ambient
CAS Number:1215738-90-1
Formula:C₉H₉ClF₃N₃OS
MDL Number:MFCD11100119