Order Entry
Netherlands
ContactUsLinkComponent
 
[5-Chloro-4-(trifluoromethyl)pyridin-2-yl]methanol 97%
Catalog # APOSPC28226-1G
Supplier:  Apollo Scientific
CAS Number:  
[5-Chloro-4-(trifluoromethyl)pyridin-2-yl]methanol 97%
Catalog # APOSPC28226-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.57 g/mol
Storage Temperature:Ambient
CAS Number:1370587-20-4
Formula:C₇H₅ClF₃NO
MDL Number:MFCD22126113