
5-Fluoro-2-iodobenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52548-63-7
:
Formula:C₇H₄FIO₂
MDL Number:MFCD00837316
:266.01 g/mol
Melting Pt:145…149 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together