
4-Fluoro-2-iodobenzoic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56096-89-0
:
Formula:C₇H₄FIO₂
MDL Number:MFCD04108278
:266.01 g/mol
Melting Pt:140…147 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together