

Copper(I) trifluoromethanesulphonate benzene complex, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:42152-46-5
:MFCD26131763
Formula:[CuOSO₂CF₃]₂.C₆H₆
MDL Number:MFCD00043300
:503.33 g/mol
Melting Pt:160 °C (dec.)
UN:3175
ADR:4.1,II
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together