
5-Methyl-4-nitro-3-trifluoromethyl-1H-pyrazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27116-80-9
:
Formula:C₅H₄F₃N₃O₂
MDL Number:MFCD00156055
:195.1 g/mol
Melting Pt:74…79 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together