Methyl-4-(2-nitro-4-(trifluoromethyl)phenoxy)benzoate
Catalog # APOSPC210062-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:341.25 g/mol
Storage Temperature:Ambient
CAS Number:297148-99-3
Formula:C₁₅H₁₀F₃NO₅
MDL Number:MFCD00245361