

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23163-86-2
:
Formula:C₈H₃ClF₃NS
MDL Number:MFCD00041090
:237.63 g/mol
Boiling Pt:248…251 °C
Melting Pt:34…36 °C
Density:1.47 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together