Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate 98%
Corrosive
Irritant
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate 98%
Catalog # APOSPC2074NK-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate 98%
Catalog # APOSPC2074NK-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.63 g/mol
Boiling Pt:239 °C
Density:1.45 g/cm³ (25 °C)
Flash Pt:>110 °C
UN:2922
ADR:8+6.1,III
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:23165-49-3
Formula:C₈H₃ClF₃NS
MDL Number:MFCD00041055