

4-Chloro-2-(trifluoromethyl)phenyl isocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16588-69-5
:
Formula:C₈H₃ClF₃NO
MDL Number:MFCD00013873
:221.57 g/mol
Boiling Pt:114…115 °C/10mmHg
Density:1.467 g/cm³ (25 °C)
Flash Pt:94 °C
Storage Temperature:Refrigerator
UN:2206
ADR:6.1,III
:GHS07
:GHS08
Frequently Bought Together