

2-Chloro-5-(trifluoromethyl)phenyl isocyanate 98%
Catalog # APOSPC2074NE-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.57 g/mol
Boiling Pt:63…64 °C/5mmHg
Density:1.48 g/cm³ (25 °C)
Flash Pt:92 °C
UN:2206
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Refrigerator
CAS Number:50528-86-4
Formula:C₈H₃ClF₃NO
MDL Number:MFCD00037029