N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57292-45-2
:
Formula:C₁₄H₁₈FNO₄
MDL Number:MFCD00079669
Melting Pt:83…86 °C
:283.30 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together