Order Entry
Netherlands
ContactUsLinkComponent
2-(Trifluoromethyl)pyrimidin-5-amine 98%
Irritant
2-(Trifluoromethyl)pyrimidin-5-amine 98%
Catalog # APOSPC201294-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Trifluoromethyl)pyrimidin-5-amine 98%
Catalog # APOSPC201294-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.1000 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:73418-87-8
Formula:C₅H₄F₃N₃
MDL Number:MFCD09754047