3-Bromo-7-(trifluoromethyl)imidazo[1,2-a]pyrimidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:375857-65-1
:
Formula:C7H3BrF3N3
MDL Number:MFCD08234995
:266.0180 g/mol
Storage Temperature:Ambient
Frequently Bought Together