Methyl-4-fluoro-3-(trifluoromethyl)benzoate 97+%
Catalog # APOSPC201175-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:222.1400 g/mol
Storage Temperature:Ambient
CAS Number:176694-36-3
Formula:C₉H₆F₄O₂
MDL Number:MFCD06203709