
5-(Trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Catalog # APOSPC200523-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:180.08 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:543739-84-0
:1259932-11-0
Formula:C₅H₃F₃N₂O₂
MDL Number:MFCD04125015