N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide
Catalog # APOSPC200462-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:282.65 g/mol
Storage Temperature:Ambient
CAS Number:1610047-55-6
Formula:C₁₀H₁₀ClF₃N₂O₂
MDL Number:MFCD28023503