
Methyl-2-cyano-4-fluoro-5-methoxybenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1007455-23-3
:
Formula:C₁₀H₈FNO₃
MDL Number:MFCD20482373
:209.18 g/mol
Melting Pt:100…102 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together