
Methyl-4-fluoro-3-methoxybenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74385-37-8
:
Formula:C₉H₉FO₃
MDL Number:MFCD06203732
:184.17 g/mol
Melting Pt:69…71 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together