3-Chloro-2-(piperidin-3-yloxy)-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1242268-26-3
:
Formula:C₁₁H₁₂ClF₃N₂O
MDL Number:MFCD16710270
:280.68 g/mol
Storage Temperature:Ambient
Frequently Bought Together