
2-Bromo-6-(trifluoromethyl)nicotinic acid
Catalog # APOSPC200239-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:130…132 °C
:270.01 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:749875-07-8
Formula:C₇H₃BrF₃NO₂
MDL Number:MFCD09909645