1-[3-(Trifluoromethyl)pyridin-2-yl]ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:749257-78-1
:
Formula:C₈H₆F₃NO
MDL Number:MFCD11044107
:189.14 g/mol
Melting Pt:65…70 °C
Storage Temperature:Ambient
Frequently Bought Together