
5-(Trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:129768-28-1
:
Formula:C₅H₃F₃N₂O₂
MDL Number:MFCD03094951
:180.09 g/mol
Melting Pt:171…173 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together