
3-Nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1186404-88-5
:
Formula:C₈H₄F₃N₃O₂
MDL Number:MFCD12827812
:231.14 g/mol
Melting Pt:118…119 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together