2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-1,3-benzodiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1393845-65-2
:
Formula:C₁₃H₇ClF₃N₃
MDL Number:MFCD22383952
:297.67 g/mol
Storage Temperature:Ambient
Frequently Bought Together