Order Entry
Netherlands
ContactUsLinkComponent
1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Irritant
1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC1969-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC1969-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1017793-88-2
Formula:C₇H₆F₃N₃O₃
MDL Number:MFCD01320690