
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22123-14-4
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00052900
:195.57 g/mol
Boiling Pt:91…93 °C/69mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together