
2-Chloro-2,2-dfluoroacetyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:354-24-5
:
Formula:C₂Cl₂F₂O
MDL Number:MFCD00039317
Boiling Pt:49…50 °C
Storage Temperature:Refrigerator
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together