
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline 95%
Catalog # APOSPC1486-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:118…124 °C
:324.53 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:123637-51-4
Formula:C₁₁H₆BrClF₃N
MDL Number:MFCD00153079