1-Bromo-2-fluorobenzene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1072-85-1
:
Formula:C₆H₄BrF
MDL Number:MFCD00000282
:175 g/mol
Boiling Pt:155…157 °C
Melting Pt:–8 °C
Density:1.601 g/cm³ (25 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:214-018-3
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together