
2-Bromo-6-chloro-4-(trifluoromethyl)aniline 98%
Catalog # APOSPC1374-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.47 g/mol
Melting Pt:27…30 °C
Flash Pt:110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:109919-26-8
Formula:C₇H₄BrClF₃N
MDL Number:MFCD00042180