Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Irritant
3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Catalog # APOSPC11262-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Catalog # APOSPC11262-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:59108-47-3
Formula:C₁₀H₅BrF₃NO
MDL Number:MFCD03407378