
Potassium(2-methylphenyl)trifluoroborate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:274257-34-0
:
Formula:C₇H₇BF₃K
MDL Number:MFCD03790006
:198.04 g/mol
Melting Pt:90…94 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together