

4'-Trifluoromethylbiphenyl-4-carbaldehyde
Catalog # APOSPC11172-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.2200 g/mol
Melting Pt:70…72 °C
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:90035-34-0
Formula:C₁₄H₉F₃O
MDL Number:MFCD01862519