
4-Fluoro-3-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2357-47-3
:
Formula:C₇H₅F₄N
MDL Number:MFCD00007834
:179.12 g/mol
Boiling Pt:207…208 °C
Density:1.393 g/cm³ (25 °C)
Flash Pt:92 °C
EINECS:219-095-7
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together