

2-Chloro-4-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39885-50-2
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00042563
:195.57 g/mol
Boiling Pt:214…218 °C
Density:1.434 g/cm³ (25 °C)
Flash Pt:103 °C
Storage Temperature:Refrigerator
EINECS:254-674-8
UN:2811
ADR:6.1,III
:GHS07
:GHS09
Frequently Bought Together