
2-Bromo-6-nitro-4-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113170-71-1
:
Formula:C₇H₄BrF₃N₂O₂
MDL Number:MFCD00042151
:285.02 g/mol
Melting Pt:69…74 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together