Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-4-(trifluoromethyl)aniline 98%
2-Bromo-4-(trifluoromethyl)aniline 98%
  APOSPC1045-100G
 :  
2-Bromo-4-(trifluoromethyl)aniline 98%
  APOSPC1045-100G
 :  
  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401
 :
 :110 °C/10mmHg
 :26…28 °C
 :1.7 g/cm³ (25 °C)
 :>110 °C
 :Refrigerator
 :261-035-7
 :2811
 :6.1,III
 :240.02 g/mol
 :GHS07
 :57946-63-1
 :C₇H₅BrF₃N
 :MFCD00042150