Order Entry
Netherlands
ContactUsLinkComponent
2-Fluoro-5-nitro-4-picoline 97%
Flammable
Irritant
2-Fluoro-5-nitro-4-picoline 97%
Catalog # APOSPC10419-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
2-Fluoro-5-nitro-4-picoline 97%
Catalog # APOSPC10419-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :156.12 g/mol
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:19346-47-5
Formula:C₆H₅FN₂O₂
MDL Number:MFCD03095070