Ga naar Rubriek
Ga naar Hoofdinhoud
Ga naar Voettekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
APOSPC10373-250MG
:
Apollo Scientific
:
5-Methyl-3-(trifluoromethyl)-1H-1,2,4-triazole
APOSPC10373-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
151.09 g/mol
:
143…145 °C
:
GHS07
:
Ambient
:
667873-25-8
:
C₄H₄F₃N₃
:
MFCD03463854