
5-(2-Thienyl)-3-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:128228-96-6
:
Formula:C₈H₅F₃N₂S
MDL Number:MFCD00459838
Melting Pt:120…122 °C
:218.2 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together