Order Entry
Netherlands
ContactUsLinkComponent
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate
Irritant
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate
Catalog # APOSPC10137-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate
Catalog # APOSPC10137-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:60…61 °C
 :222.167 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:111493-74-4
Formula:C₈H₉F₃N₂O₂
MDL Number:MFCD02083398