
3-Chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-5-(trifluoromethyl)-2(1H)-pyridinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96741-18-3
:
Formula:C₁₂H₄Cl₂F₆N₂O
MDL Number:MFCD01568202
Melting Pt:107…109 °C
:377.07 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together