1-Fluoropropane 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Propyl fluoride
CAS Number:460-13-9
:
Formula:C₃H₇F
MDL Number:MFCD01320734
Boiling Pt:–3 °C
Melting Pt:–159 °C
Density:0.781 g/cm³ (25 °C)
Molecular Weight:62.09 g/mol
Flash Pt:<21 °C
Storage Temperature:Ambient
UN:3161
ADR:2.1
:GHS02
:GHS04
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together