2,3,5,6-Tetrafluoro-p-xylene 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3,5,6-Tetrafluoro-1,4-dimethylbenzene
CAS Number:703-87-7
:
Formula:C₈H₆F₄
MDL Number:MFCD00012231
Boiling Pt:143…144 °C
Molecular Weight:178.13 g/mol
Melting Pt:35…37 °C
Flash Pt:47 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together