
3-Chloro-4-(trifluoromethyl)aniline 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:445-13-6
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00278479
:195.57 g/mol
Boiling Pt:200…202 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together