
2-Bromo-4,5-bis(trifluoromethyl)aniline
Catalog # APOSPC0295-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:308.02 g/mol
Boiling Pt:117…118 °C/12mmHg
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:230295-15-5
Formula:C₈H₄BrF₆N