Order Entry
Netherlands
ContactUsLinkComponent
2,3-Difluoro-p-cresol
Corrosive
Irritant
2,3-Difluoro-p-cresol
Catalog # APOSPC0259-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2,3-Difluoro-p-cresol
Catalog # APOSPC0259-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.12 g/mol
Melting Pt:35…38 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:261763-45-5
Formula:C₇H₆F₂O
MDL Number:MFCD01631651