
2-Hydroxy-4-methoxybenzenecarbohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:41697-08-9
:
Formula:C₈H₁₀N₂O₃
MDL Number:MFCD06797386
:182.18 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together